Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGRP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173983.2 | NP_776408.2 | 1 - 2 | 131 | 129 | |
GenBank mRNA | AJ002025.1 | - | 1 - 2 | 131 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Bos taurus Probe spans exons |