Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FTL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174792.4 | NP_777217.1 | 2 - 3 | 501 | 68 | |
GenBank mRNA | AB003094.1 | - | 2 - 3 | 321 | 68 | |
AY528246.1 | - | 1 | 438 | 68 | ||
BC103021.1 | - | 1 | 450 | 68 | ||
EE236814.1 | - | 2 - 3 | 502 | 68 | ||
EV670183.1 | - | 2 - 3 | 454 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |