Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175776.2 | NP_786970.1 | 3 - 4 | 385 | 58 | |
GenBank mRNA | BC102933.1 | - | 3 - 4 | 385 | 58 | |
M64520.1 | - | 3 - 4 | 385 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |