Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014960.1 | NP_001014960.1 | 2 - 3 | 162 | 70 | |
GenBank mRNA | BT021537.1 | - | 2 - 3 | 162 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |