Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARMC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015594.3 | NP_001015594.2 | 5 - 6 | 811 | 71 | |
GenBank mRNA | BC103407.1 | - | 5 - 6 | 812 | 71 | |
BT020641.1 | - | 5 - 6 | 762 | 71 | ||
EV727326.1 | - | 2 - 3 | 547 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |