Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034780.1 | NP_001029952.1 | 5 - 6 | 544 | 117 | |
GenBank mRNA | BT020923.1 | - | 5 - 6 | 544 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Bos taurus Probe spans exons |