Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUDT21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035331.2 | NP_001030408.1 | 3 - 4 | 506 | 94 | |
GenBank mRNA | BC102697.1 | - | 3 - 4 | 507 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |