Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSD17B10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174334.3 | NP_776759.1 | 5 - 6 | 614 | 68 | |
GenBank mRNA | AB002156.1 | - | 5 - 6 | 615 | 68 | |
BC110264.1 | - | 5 - 6 | 612 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |