Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034437.1 | NP_001029609.1 | 2 - 3 | 264 | 79 | |
GenBank mRNA | BC102666.1 | - | 2 - 3 | 264 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |