Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174345.4 | NP_776770.2 | 1 - 2 | 72 | 83 | |
GenBank mRNA | BC105182.1 | - | 1 - 2 | 73 | 83 | |
BT030487.1 | - | 1 - 2 | 58 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Inventoried Probe spans exons |