Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174731.4 | NP_777156.1 | 4 - 5 | 384 | 69 | |
GenBank mRNA | AF520959.1 | - | 4 - 5 | 307 | 69 | |
AY911375.1 | - | 4 - 5 | 229 | 69 | ||
BC102873.1 | - | 4 - 5 | 385 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |