Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C25H16orf71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046621.2 | NP_001040086.1 | 4 - 5 | 851 | 77 | |
GenBank mRNA | BC112526.1 | - | 4 - 5 | 852 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |