Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PENK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174141.2 | NP_776566.1 | 1 - 2 | 168 | 80 | |
GenBank mRNA | BC111279.1 | - | 1 - 2 | 168 | 80 | |
V00109.1 | - | 105 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |