Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045875.1 | NP_001039340.1 | 1 - 2 | 315 | 80 | |
GenBank mRNA | AY924307.1 | - | 1 - 2 | 315 | 80 | |
BC118328.1 | - | 1 - 2 | 563 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |