Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046528.1 | NP_001039993.1 | 7 - 8 | 650 | 133 | |
GenBank mRNA | BC113316.1 | - | 7 - 8 | 650 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |