Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTARC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076380.2 | NP_001069848.1 | 1 - 2 | 331 | 126 | |
GenBank mRNA | BC116001.1 | - | 1 - 2 | 332 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |