Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PRXL2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076436.2 | NP_001069904.1 | 3 - 4 | 369 | 148 | |
GenBank mRNA | BC118425.1 | - | 3 - 4 | 371 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |