Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PEMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182989.3 | NP_892034.1 | 5 - 6 | 675 | 85 | |
GenBank mRNA | AY334572.1 | - | 3 - 4 | 259 | 85 | |
BC105466.1 | - | 5 - 6 | 676 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |