Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VWA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075206.1 | NP_001068674.1 | 12 - 13 | 1994 | 90 | |
GenBank mRNA | BT026224.1 | - | 12 - 13 | 1994 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Bos taurus Probe spans exons |