Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075535.2 | NP_001069003.1 | 6 - 7 | 912 | 97 | |
GenBank mRNA | BC120023.1 | - | 6 - 7 | 913 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |