Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001075815.1 | NP_001069283.1 | 6 - 7 | 1232 | 60 | |
GenBank mRNA | BC119954.1 | - | 6 - 7 | 1232 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |