Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAAF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076024.3 | NP_001069492.2 | 1 - 2 | 2062 | 100 | |
GenBank mRNA | BC120319.1 | - | 1 - 2 | 616 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |