Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UTP23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076039.2 | NP_001069507.2 | 1 - 2 | 281 | 72 | |
GenBank mRNA | BC123565.1 | - | 1 - 2 | 268 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |