Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMLHE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076064.1 | NP_001069532.1 | 3 - 4 | 477 | 117 | |
GenBank mRNA | BC120318.1 | - | 3 - 4 | 477 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |