Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KRT75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076917.1 | NP_001070385.1 | 4 - 5 | 916 | 65 | |
GenBank mRNA | BC123881.1 | - | 4 - 5 | 916 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |