Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024561.2 | NP_001019732.1 | 5 - 6 | 649 | 62 | |
GenBank mRNA | BC133593.1 | - | 5 - 6 | 649 | 62 | |
BT020721.1 | - | 4 - 5 | 555 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |