Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAG8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176619.3 | NP_788792.2 | 6 - 7 | 806 | 55 | |
GenBank mRNA | AF020510.2 | - | 6 - 7 | 762 | 55 | |
BC133476.1 | - | 6 - 7 | 807 | 55 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |