Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDK9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014935.2 | NP_001014935.2 | 4 - 5 | 463 | 69 | |
GenBank mRNA | BC118194.1 | - | 4 - 5 | 463 | 69 | |
BT020657.1 | - | 4 - 5 | 462 | 69 | ||
BT021882.1 | - | 4 - 5 | 491 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |