Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098055.1 | NP_001091524.1 | 4 - 5 | 945 | 91 | |
GenBank mRNA | BC140508.1 | - | 4 - 5 | 945 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |