Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CDKN1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100346.1 | NP_001093816.1 | 2 - 3 | 1115 | 145 | |
GenBank mRNA | BC148067.1 | - | 2 - 3 | 1115 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |