Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ACSM2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001101952.2 | NP_001095422.1 | 11 - 12 | 1543 | 72 | |
GenBank mRNA | BC151513.1 | - | 11 - 12 | 1544 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |