Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYP2C19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109792.2 | NP_001103262.1 | 6 - 7 | 982 | 67 | |
GenBank mRNA | AY265992.1 | - | 2 - 3 | 210 | 67 | |
BC153845.1 | - | 6 - 7 | 984 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |