Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099073.2 | NP_001092543.1 | 6 - 7 | 700 | 99 | |
GenBank mRNA | BC142073.1 | - | 6 - 7 | 702 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |