Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MNAT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105409.1 | NP_001098879.1 | 1 - 2 | 210 | 83 | |
GenBank mRNA | BC104522.1 | - | 1 - 2 | 210 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |