Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SHISA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145203.1 | NP_001138675.2 | 3 - 4 | 1027 | 68 | |
GenBank mRNA | EH181025.1 | - | 3 - 4 | 363 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Bos taurus Probe spans exons |