Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTNL9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001103345.1 | NP_001096815.1 | 6 - 7 | 1069 | 76 | |
GenBank mRNA | BC149479.1 | - | 6 - 7 | 1069 | 76 | |
BT029803.1 | - | 6 - 7 | 1090 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |