Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001205447.1 | NP_001192376.1 | 2 - 3 | 2126 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |