Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BSPRY | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206723.1 | NP_001193652.1 | 4 - 5 | 555 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |