Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FEM1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001192383.2 | NP_001179312.1 | 1 - 2 | 363 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Bos taurus Probe spans exons |