Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FGF7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193131.1 | NP_001180060.1 | 2 - 3 | 392 | 138 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Bos taurus Probe spans exons |