Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aak-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029697.5 | NP_001024868.1 | 2 - 3 | 639 | 66 | |
NM_078309.7 | NP_510710.2 | 3 - 4 | 782 | 66 | ||
NM_078310.7 | NP_510711.2 | 3 - 4 | 784 | 66 | ||
GenBank mRNA | AY347273.1 | - | 3 - 4 | 782 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |