Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hrpf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170888.6 | NP_740877.1 | 4 - 5 | 1051 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |