Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hmr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026479.4 | NP_001021650.1 | 25 - 26 | 6053 | 88 | |
NM_001264632.3 | NP_001251561.1 | 2 - 3 | 377 | 88 | ||
NM_001381215.1 | NP_001366962.1 | 5 - 6 | 1218 | 88 | ||
GenBank mRNA | AJ307058.1 | - | 25 - 26 | 6053 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |