Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ced-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026601.6 | NP_001021772.1 | 3 - 4 | 453 | 62 | |
NM_001383734.1 | 4 - 5 | 588 | 62 | |||
NM_170927.5 | NP_740922.1 | 3 - 4 | 453 | 62 | ||
GenBank mRNA | AF332568.1 | - | 3 - 4 | 447 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Caenorhabditis elegans Probe spans exons |