Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ekl-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_170949.7 | NP_740945.1 | 6 - 7 | 1474 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Caenorhabditis elegans Probe spans exons |