Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-87 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025921.4 | NP_001021092.1 | 1 - 2 | 725 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |