Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sem-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059596.5 | NP_491997.1 | 1 - 2 | 78 | 114 | |
GenBank mRNA | U60112.1 | - | 1 - 2 | 78 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |