Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prpf-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001263462.4 | NP_001250391.1 | 3 - 4 | 935 | 103 | |
NM_001263463.4 | NP_001250392.1 | 2 - 3 | 803 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |