Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dylt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_059662.7 | NP_492063.1 | 2 - 3 | 272 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |