Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rskn-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047180.4 | NP_001040645.1 | 2 - 3 | 619 | 89 | |
NM_001361775.4 | NP_001348666.1 | 2 - 3 | 621 | 89 | ||
NM_001361776.3 | NP_001348667.1 | 2 - 3 | 619 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |